missing translation for 'onlineSavingsMsg'
Learn More

JW 642, MedChemExpress

Product Code. 16455098
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16455098 10 mg 10mg
16421337 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16455098 Supplier Medchem Express Supplier No. HY12332
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress JW 642 is a potent inhibitor of monoacylglycerol lipase (MAGL) that displays IC50 values of 7.6, 14, and 3.7 nM for inhibition of MAGL in mouse, rat, and human brain membranes, respectively.

JW 642 is a potent inhibitor of monoacylglycerol lipase (MAGL) that displays IC50 values of 7.6, 14, and 3.7 nM for inhibition of MAGL in mouse, rat, and human brain membranes, respectively.

Specifications

Chemical Name or Material JW 642
CAS 1416133-89-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H20F6N2O3
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (216.27 mM)
SMILES O=C(OC(C(F)(F)F)C(F)(F)F)N(CC1)CCN1CC2=CC(OC3=CC=CC=C3)=CC=C2
Molecular Weight (g/mol) 462.39
Formula Weight 462.39
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.