missing translation for 'onlineSavingsMsg'
Learn More

JW74, MedChemExpress

Product Code. 16490338
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490338 5 mg 5mg
16457878 10 mg 10mg
16433827 50 mg 50mg
16413686 25 mg 25mg
16407696 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16490338 Supplier Medchem Express Supplier No. HY19739
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress JW74 antagonizes LiCl-induced activation of the canonical Wnt signaling with an IC50 of 420 nM.

JW74 antagonizes LiCl-induced activation of the canonical Wnt signaling with an IC50 of 420 nM.

Specifications

Chemical Name or Material JW74
CAS 863405-60-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H20N6O2S
Quantity 5 mg
Solubility Information DMSO : ≥ 50 mg/mL (109.52 mM)
SMILES CC1=CC=C(C2=NOC(CSC3=NN=C(C4=CC=NC=C4)N3C5=CC=C(OC)C=C5)=N2)C=C1
Molecular Weight (g/mol) 456.52
Formula Weight 456.52
Percent Purity 98.32%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.