missing translation for 'onlineSavingsMsg'
Learn More

K-Ras G12C-IN-3, MedChemExpress

Product Code. 16437486
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16437486 100 mg 100mg
16497528 10 mg 10mg
16431739 1 mg 1mg
16413557 50 mg 50mg
16410018 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16437486 Supplier Medchem Express Supplier No. HY18606
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress K-Ras G12C-IN-3 is a novel and irreversible inhibitor of mutant K-ras G12C.

K-Ras G12C-IN-3 is a novel and irreversible inhibitor of mutant K-ras G12C.

Specifications

Chemical Name or Material K-Ras G12C-IN-3
CAS 1629268-19-4
Color White
Purity Grade Notes Research
Molecular Formula C21H19Cl3N2O3
Quantity 100 mg
Solubility Information DMSO : ≥ 30 mg/mL (66.12 mM)
SMILES ClC(C=C(OC)C(C(N1CCN(C(C=C)=O)CC1)=O)=C2)=C2C3=CC(Cl)=CC=C3Cl
Molecular Weight (g/mol) 453.75
Formula Weight 453.75
Percent Purity 97.18%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.