missing translation for 'onlineSavingsMsg'
Learn More
Learn More
K-Ras G12C-IN-3, MedChemExpress
Description
K-Ras G12C-IN-3 is a novel and irreversible inhibitor of mutant K-ras G12C.
Specifications
Specifications
| Chemical Name or Material | K-Ras G12C-IN-3 |
| CAS | 1629268-19-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H19Cl3N2O3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 30 mg/mL (66.12 mM) |
| SMILES | ClC(C=C(OC)C(C(N1CCN(C(C=C)=O)CC1)=O)=C2)=C2C3=CC(Cl)=CC=C3Cl |
| Molecular Weight (g/mol) | 453.75 |
| Formula Weight | 453.75 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?