missing translation for 'onlineSavingsMsg'
Learn More

Kaempferitrin, TRC

Product Code. 17949654
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17949654 5 mg 5mg
17918314 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17949654 Supplier Toronto Research Chemicals Supplier No. TRCK1001005MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 482-38-2
Molecular Formula C27 H30 O14
Molecular Weight (g/mol) 578.52
Synonym 4H-1-Benzopyran-4-one, 3,7-bis[(6-deoxy-α-L-mannopyranosyl)oxy]-5-hydroxy-2-(4-hydroxyphenyl)-, Kaempferitrin (6CI,7CI,8CI), 3,4',5,7-Tetrahydroxyflavone 3,7-dirhamnoside, 3,7-Bis[(6-deoxy-α-L-mannopyranosyl)oxy]-5-hydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Kaempferol 3,7-O-α-L-dirhamnoside, Kaempferol 3,7-bisrhamnoside, Kaempferol 3,7-di-O-rhamnopyranoside, Kaempferol 3,7-di-O-rhamnoside, Kaempferol 3,7-di-O-α-L-rhamnopyranoside, Kaempferol 3,7-di-α-L-rhamnopyranoside, Kaempferol 3,7-di-α-L-rhamnoside, Kaempferol 3,7-dirhamnoside, Kaempferol 3-O-rhamnoside 7-O-rhamnoside, Kaempferol 3-O-α-L-rhamnopyranoside 7-O-α-L-rhamnopyranoside, Kaempferol-3,7-O-dirhamnose, Lespedin, Lespenephryl
IUPAC Name 5-hydroxy-2-(4-hydroxyphenyl)-3,7-bis[[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy]chromen-4-one
SMILES C[C@@H]1O[C@@H](OC2=C(Oc3cc(O[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)cc(O)c3C2=O)c5ccc(O)cc5)[C@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C27H30O14/c1-9-17(30)20(33)22(35)26(37-9)39-13-7-14(29)16-15(8-13)40-24(11-3-5-12(28)6-4-11)25(19(16)32)41-27-23(36)21(34)18(31)10(2)38-27/h3-10,17-18,20-23,26-31,33-36H,1-2H3/t9-,10-,17-,18-,20+,21+,22+,23+,26-,27-/m0/s1
Formula Weight 578.1636
Chemical Name or Material Kaempferitrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.