missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Kaempferol, 97%

Product Code. 10872631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10872631 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10872631 Supplier Thermo Scientific Chemicals Supplier No. 434731000
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 520-18-3 | C15H10O6 | 286.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 520-18-3
Melting Point 282°C to 286°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C15H10O6
MDL Number MFCD00016938
Quantity 100 mg
Merck Index 15,5322
Synonym kaempferol, kaempherol, kempferol, populnetin, rhamnolutein, robigenin, trifolitin, pelargidenolon, rhamnolutin, swartziol
InChI Key IYRMWMYZSQPJKC-UHFFFAOYSA-N
SMILES OC1=CC=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1
IUPAC Name 3,5,7-trihydroxy-2-(4-hydroxyphenyl)chromen-4-one
Molecular Weight (g/mol) 286.24
PubChem CID 5280863
ChEBI CHEBI:28499
Formula Weight 286.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Kaempferol
Show More Show Less
compliance-icons
Product Identifier
  • Kaempferol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.