Learn More
Kallikrein 3/PSA Antibody (CL9414) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-05505AF647
Product Code. 30290165
  • 737.00 € / 0.10mL
Description
Kallikrein 3/PSA Monoclonal antibody specifically detects Kallikrein 3/PSA in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kallikrein 3/PSA
Monoclonal
Alexa Fluor 647
APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin
This antibody was developed using a recombinant protein derived from P07288, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cellular Markers, Prostate Cancer, Tumor Biomarkers
354
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL9414
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified