missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 7 Antibody (KLK7/4692), Novus Biologicals™
Mouse Monoclonal Antibody
249.00 € - 494.00 €
Specifications
| Antigen | Kallikrein 7 |
|---|---|
| Clone | KLK7/4692 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680295
|
Novus Biologicals
NBP3-20331-100ug |
100 μg |
494.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18612345
|
Novus Biologicals
NBP3-20331-20ug |
20 μg |
249.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kallikrein 7 Monoclonal antibody specifically detects Kallikrein 7 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Kallikrein 7 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 10mM PBS with 0.05% BSA | |
| 5650 | |
| IgG2b κ | |
| Protein A or G purified |
| KLK7/4692 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.4.21, EC 3.4.21.117, hK7, hSCCE, kallikrein 7 (chymotryptic, stratum corneum), kallikrein-related peptidase 7, protease, serine, 6, PRSS6, SCCEkallikrein-7, Serine protease 6, signal protein, Stratum corneum chymotryptic enzyme | |
| Recombinant fragment of human Kallikrein 7 protein (around aa 1-200) (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title