missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kallikrein 9 Monoclonal Antibody (366721)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561315
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. KLK9 is a novel kallikrein with potential application for diagnosis, monitoring and therapeutics of various cancers including those of the breast, prostate and testis.
Specifications
| Kallikrein 9 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9UKQ9 | |
| KLK9 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 9, Asp20-Asn250. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| 366721 | |
| Unconjugated | |
| KLK9 | |
| 1200016C12Rik; AI324041; kallikrein 9; kallikrein related peptidase 9; kallikrein related-peptidase 9; kallikrein-9; Kallikrein-like protein 3; kallikrein-related peptidase 9; KLK9; KLK-9; KLKL3; KLK-L3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 284366 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction