missing translation for 'onlineSavingsMsg'
Learn More

Melford Kanamycin A

Product Code. 30018363
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018363 1 g 1g
30018364 10 g 10g
30018365 25 g 25g
30018366 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30018363 Supplier Melford Supplier No. K220001.0
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Kanamycin A is an aminoglycoside antibiotic that is widely used in the field of life sciences.

Kanamycin A is an aminoglycoside antibiotic that is widely used in the field of life sciences. It is derived from the bacterium Streptomyces kanamyceticus and has potent antimicrobial properties against a broad spectrum of bacteria.

In the life science marketplace, Kanamycin A is primarily used as a selective agent in microbiology and molecular biology applications. It is commonly incorporated into culture media to inhibit the growth of bacteria that do not possess the kanamycin resistance gene. This selective property allows for the isolation and identification of cells or organisms that have been genetically modified to express the resistance gene.

Researchers and scientists working in various fields such as molecular biology, genetic engineering, and biotechnology utilize Kanamycin A in experiments involving recombinant DNA technology.

It is often employed as a selectable marker in plasmids and other vectors to ensure that only cells carrying the desired genetic material survive and proliferate.

Additionally, Kanamycin A is used in laboratory settings to study bacterial resistance mechanisms and evaluate the efficacy of antimicrobial compounds. Its broad-spectrum activity and low toxicity make it a valuable tool for researchers studying bacterial genetics, protein expression, and antibiotic resistance.

Specifications

Chemical Name or Material Kanamycin A
CAS 25389-94-0
Quantity 1 g
Molecular Formula C18H36N4O11-H2SO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.