missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Kanamycin Acid Sulfate ReadyMade™ Solution

Product Code. 30044058
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044058 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044058 Supplier TOKU-E™ Supplier No. K03510x1ML
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kanamycin Acid Sulfate ReadyMade™ Solution is provided as a sterile-filtered solution of Kanamycin Acid Sulfate in water.

  • CAS Number: 64013-70-3
  • Molecular Formula: C18H36N4O11 · 2 H2SO4
  • Molecular Weight: 680.65 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Streptomycines kanamyceticus
  • Spectrum: Kanamycin is a broad spectrum and inhibits growth of Gram-positive bacteria, Gram-negative bacteria, and mycoplasmas
  • Mechanism of Action: Aminoglycosides target the 30S ribosomal subunit, lodging between the 16S rRNA and S12 protein. This interferes with the translational initiation complex causing misreading of the mRNA, and production of a faulty or non-existent protein

Specifications

CAS 64013-70-3
Chemical Name or Material Kanamycin Acid Sulfate
Color Clear, colorless
Content And Storage -20°C, Protect from light
Description Kanamycin Acid Sulfate ReadyMade™ Solution 50 mg/mL in Water
Concentration 50.0 to 52.5 mg/mL
Form Clear, colorless solution
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid in Water
For Use With (Application) Microbiology, Plant Biology, Eukaryotic Cell Culture, Cancer Application
Product Line ReadyMade™ Solutions
Quantity 10 x 1 mL
Shipping Condition Ice Pack
Source Streptomyces Kanamyceticus
Species Bacteria
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.