missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin Solution, MP Biomedicals

Product Code. 11400435 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400435 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11400435 Supplier MP Biomedicals Supplier No. 091672048
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibiotic solution Concentration: 5mg/mL Effective against mycoplasma, gram-positive and gram-negative bacteria

Specifications

Type Kanamycin
Physical Form Liquid
CAS 25389-94-0, 7732-18-5
Quantity 50 mL
Grade Not Available
Chemical Name or Material Kanamycin Solution
compliance-icons
Product Identifier
  • Kanamycin Monosulfate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1A
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Kanamycin Monosulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.