missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Kanamycin Sulfate
GREENER_CHOICE

Product Code. 11578876
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11578876 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11578876 Supplier Gibco™ Supplier No. 15160047
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Kanamycin Sulfate

Kanamycin Sulfate is a water-soluble antibiotic originally purified from the bacterium Streptomyces kanamyceticus. Kanamycin acts by binding to the 30S subunit of the bacterial ribosome and inhibiting protein synthesis in susceptible bacteria. Gibco™ Kanamycin is effective against a wide variety of gram-negative and some gram-positive bacteria, and is used for the prevention of bacterial contamination of cell cultures. This product is provided as a 10 mg/ml stock solution. The recommended working concentration for cell culture is 100μg/ml. We offer a variety of antibiotics and antimycotics for cell culture applications.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Dual-Site cGMP Manufacturing
Gibco™ MEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ MEM product made in our Grand Island facility (15160-054). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 13485, cGMP, FDA
Content And Storage –5 to –20°C
Description Kanamycin Sulfate, 100mL, 10mg/mL, 100X Concentration, –5 to –20°C Storage Temperature, Antibiotic (Selective), Kanamycin Agent, Antibiotic (Selective) Reagent Type, 12 Months Shelf Life, ISO 13485/cGMP/FDA Certified, Dry Ice/Frozen
Concentration 100 X
Form Liquid
Product Type Antibiotic
For Use With (Application) Transformation, bacterial selection
Quantity 100 mL
compliance-icons
Product Identifier
  • Kanamycin sulphate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Kanamycin sulphate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.