missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin sulfate, MedChemExpress

Product Code. 16471619
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471619 1 g 1g
16419627 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471619 Supplier Medchem Express Supplier No. HY16566A
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Kanamycin (Kanamycin A) sulfate is an orally active antibacterial (gram-negative/positive bacteria) agent, inhibits translocation and causes misencoding by binding to the 70 S ribosomal subunit. Kanamycin sulfate shows good inhibitory activity to both M. tuberculosis (sensitive and drug-resistant ) and K. pneumonia, which can be used in studies of tuberculosis and pneumonia.

Kanamycin (Kanamycin A) sulfate is an orally active antibacterial (gram-negative/positive bacteria) agent, inhibits translocation and causes misencoding by binding to the 70 S ribosomal subunit. Kanamycin sulfate shows good inhibitory activity to both M. tuberculosis (sensitive and drug-resistant ) and K. pneumonia, which can be used in studies of tuberculosis and pneumonia.

Specifications

Chemical Name or Material Kanamycin sulfate
Physical Form Solid
CAS 25389-94-0
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C18H38N4O15S
For Use With (Application) COVID-19-immunoregulation
Synonym Kanamycin A sulfate
Solubility Information H2O : 31.25 mg/mL (53.64 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 1 mg/mL (1.72 mM; ultrasonic and warming and heat to 80°C)
SMILES OS(O)(=O)=O.O[C@H]1[C@](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@@]3([H])O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)([H])O[C@H](CN)[C@@H](O)[C@@H]1O
Molecular Weight (g/mol) 582.58
Formula Weight 582.58
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.