missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin sulfate from Streptomyces kanamycetikus, MP Biomedicals™

Product Code. 11490276 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490276 5 g 5g
11400056 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490276 Supplier MP Biomedicals Supplier No. 0219985305
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kanamycin sulfate from Streptomyces kanamycetikus USP Grade, White Powder

  • It is a broad spectrum aminoglycoside-antibiotic derived from Streptomyces kanamyceticus.
  • It is used as an additive in culture media for the isolation of group D streptococci on Kanamycin Esculin Azide Agar and for selection of transformed plant cells containing the neomycin phosphotransferase on a kanamycin-medium.
  • Kanamycin sulfate can also be used as a selection agent for cells transformed with kanamycin B resistance gene.
  • Kanamycin sulfate is an antimicrobial agent effective against Gram-negative and Gram-positive bacteria and mycoplasma.
  • It binds to the 70S ribosomal subunit, inhibits translocation, and elicits miscoding.
  • Cross-resistance occurs between kanamycin and neomycin, framycetin, and paromomycin, and partial cross-resistance has been reported between kanamycin and streptomycin. Aminoglycosidemodifying enzymes (acetyltransferase, phosphotransferase, nucleotidyltransferase) alter the antibiotic, preventing its interaction with ribosomes.

Chemical Identifiers

CAS 25389-94-0
Molecular Formula C18H38N4O15S
Molecular Weight (g/mol) 582.58
MDL Number MFCD00070253
InChI Key OOYGSFOGFJDDHP-KMCOLRRFSA-N
Synonym Kanamycin A, Kanamycin sulfate salt

Specifications

Type Kanamycin
Physical Form Powder
pH 6.5 to 8.5 (1% solution)
Melting Point >250°C
Loss on Drying <4%
Potency >750 µg/mg
Color White
Quantity 5 g
Solubility Information Soluble in 10-50mg/mL of Water
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.