missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Kanamycin Sulphate (100X)
GREENER_CHOICE

Product Code. 11588876
Change view
Click to view available options
Quantity:
20 x 100mL
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11588876 20 x 100mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11588876 Supplier Gibco™ Supplier No. 15160070

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

20 x 100mL

Kanamycin Sulphate (100X)

Kanamycin Sulfate is a water-soluble antibiotic originally purified from the bacterium Streptomyces kanamyceticus. Kanamycin acts by binding to the 30S subunit of the bacterial ribosome and inhibiting protein synthesis in susceptible bacteria. Gibco™ Kanamycin is effective against a wide variety of gram-negative and some gram-positive bacteria, and is used for the prevention of bacterial contamination of cell cultures. This product is provided as a 10 mg/ml stock solution. The recommended working concentration for cell culture is 100μg/ml. We offer a variety of antibiotics and antimycotics for cell culture applications.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Dual-Site cGMP Manufacturing
Gibco™ Kanamycin Sulfate is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Kanamycin Sulfate product made in our Grand Island facility (15160-054). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 13485, cGMP, FDA
Content And Storage –5 to –20°C
Description Kanamycin Sulphate, 20 x 100mL, 100X Concentration, –5 to –20°C Storage Temperature, Antibiotic (Selective), Kanamycin Agent, Antibiotic (Selective) Reagent Type, 12 Months Shelf Life, ISO 13485/cGMP/FDA Certified, Frozen, Fewer Resources
Concentration 100 X
Form Liquid
Product Type Antibiotic
For Use With (Application) Transformation, bacterial selection
Quantity 20 x 100mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.