missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappa Light Chain Antibody (SPM508), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34366-0.1mg
This item is not returnable.
View return policy
Description
Kappa Light Chain Monoclonal antibody specifically detects Kappa Light Chain in Human, Rat (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Kappa Light Chain | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5-2.0 ug/million cells, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 ug/mL | |
| P01601, P01834 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat (Negative) | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| SPM508 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
| Human B-Lymphoma Cells | |
| 0.1 mg | |
| Adaptive Immunity, Immunology | |
| 3514 | |
| Store at 4C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction