missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Reagent, for Karl Fischer Analysis, Modified, Solution ≈5mg H2O/mL
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Boiling Point | 124°C |
| Color | Brown |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Packaging | Plastic Coated Glass bottle |
Safety and Handling
Product Identifier
- Karl Fischer reagent
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
- H370-Causes damage to organs.
- H372-Causes damage to organs.
- H360FD-May damage fertility. May damage the unborn child.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?