missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Reagent Solution A, for pyridine-based volumetric KF titration (pyridine, sulfur dioxide, methanol), Honeywell™ Fluka™

Product Code. 15605120 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
Product Code. Quantity unitSize
15605120 6 x 1 L Pack of 6
15695110 1 L 1 L
2 options
This item is not returnable. View return policy

Product Code. 15605120

Brand: Honeywell-Fluka 361166X1L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

for pyridine-based volumetric Karl Fischer titration (pyridine, sulfur dioxide, methanol)

  • Solutions are prepared with raw materials which meet or exceed requirements of the American Chemical Society
  • Where no ACS specifications exist, chemicals of highest purity available are used
  • All aqueous solutions are prepared using high purity, deionized, water which meets requirements for ASTM type 1 Reagent Grade Water
  • Exacting production and quality control records are maintained providing traceability of raw materials
  • All solutions are guaranteed to meet label specifications
  • All solution labels meet or exceed all ISO, DOT and OSHA regulations
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Karl Fischer Reagent Solution A
Name Note for pyridine-based volumetric KF titration (pyridine, sulfur dioxide, methanol)
Boiling Point >60°C
CAS Min % ≥10.0000%, ≥25.0000%
CAS Max % <20.0000%, <50.0000%
CAS 7446-09-5, 110-86-1
Density ∼1.01 g/mL
Quantity 6 x 1 L
Flash Point 16°C
Packaging Glass Bottle
UN Number UN1992
Grade Karl Fischer
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Karl Fischer reagent solution A
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : methanol, pyridine,sulphur dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.