missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ kb NB 142-70

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective protein kinase D (PKD) inhibitor (IC50 values are 28.3, 58.7 and 53.2 nM for PKD1, 2 and 3 respectively). More potent analog of CID 755673 (Cat. No. 3327) with 7-fold greater inhibition. Inhibits prostrate cancer cell migration and invasion and reduces wound healing in vitro; displays prominent cytotoxic and anti-proliferative effects.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 251.32 |
| Purity | >98% |
| Molecular Weight (g/mol) | 251.318 |
| Chemical Name or Material | kb NB 142-70 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction