Learn More
KCNJ10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-20149
 View more versions of this product
Product Code. 18748743
  • 509.00 € / 0.10mL
Description
KCNJ10 Polyclonal specifically detects KCNJ10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LYOPH
A synthetic peptide from amino acid region 330-379 of human KCNJ10 conjugated to blue carrier protein was used as the antigen. The antigen is homologous in rat and mouse.
RUO
KCNJ10
Unpurified
Human, Mouse, Rat