missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK3 Antibody (S374-48), Novus Biologicals™
Mouse Monoclonal Antibody
174.00 € - 561.00 €
Specifications
| Antigen | KCNK3 |
|---|---|
| Clone | S374-48 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671408
|
Novus Biologicals
NBP2-42202-0.025mg |
0.025 mg |
174.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658506
|
Novus Biologicals
NBP2-42202 |
0.1 mg |
561.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KCNK3 Monoclonal antibody specifically detects KCNK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| KCNK3 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Neuroscience | |
| PBS (pH 7.4), 50% Glycerol | |
| Acid-sensitive potassium channel protein TASK-1, cardiac potassium channel, K2p3.1, OAT1, potassium channel subfamily K member 3, potassium channel, subfamily K, member 3, potassium inwardly-rectifying channel, subfamily K, member 3, TASK1, TASK-1, TASKacid-sensitive potassium channel protein TASK, TBAK1, TWIK-related acid-sensitive K(+) channel 1, TWIK-related acid-sensitive K+ channel, two P domain potassium channel, Two pore K(+) channel KT3.1, Two pore potassium channel KT3.1 | |
| Fusion protein amino acids 251-411 (cytoplasmic C-terminus) of rat Acid-sensitive potassium channel protein TASK or TASK1. Mouse: 96% identity (156/161 amino acids identical). Human: 76% identity (163/161 amino acids identical). <30% identity with TASK3. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S374-48 | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| O54912 | |
| 3777 | |
| IgG2b | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title