missing translation for 'onlineSavingsMsg'
Learn More

KDU691, MedChemExpress

Product Code. 16418157
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16418157

Brand: Medchem Express HY12912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KDU691, an imidazopyrazine with potent anti-parasitic activity against blood stage schizonts, gametocytes and liver stages, is a Plasmodium PI4K inhibitor. KDU691 selectively inhibits dihydroartemisinin-pretreated Plasmodium falciparum ring-stage parasites.

KDU691, an imidazopyrazine with potent anti-parasitic activity against blood stage schizonts, gametocytes and liver stages, is a Plasmodium PI4K inhibitor. KDU691 selectively inhibits dihydroartemisinin-pretreated Plasmodium falciparum ring-stage parasites.

Specifications

Chemical Name or Material KDU691
Physical Form Solid
CAS 1513879-19-0
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H18ClN5O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 150 mg/mL (357.26 mM; Need ultrasonic and warming)
SMILES ClC1=CC=C(N(C(C2=CN3C(C=N2)=NC=C3C4=CC=C(C(NC)=O)C=C4)=O)C)C=C1
Molecular Weight (g/mol) 419.86
Formula Weight 419.86
Percent Purity 99.43%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.