missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KELL Monoclonal Antibody (214711)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA560984
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a type II transmembrane glycoprotein that is the highly polymorphic Kell blood group antigen. The Kell glycoprotein links via a single disulfide bond to the XK membrane protein that carries the Kx antigen. The encoded protein contains sequence and structural similarity to members of the neprilysin (M13) family of zinc endopeptidases.
Specifications
| KELL | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| Q9EQF2 | |
| KEL | |
| Mouse myeloma cell line NS0-derived recombinant mouse Kell, Phe50-Trp713. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 214711 | |
| Unconjugated | |
| KEL | |
| CD238; ECE3; Kel; Kel blood group protein; Kell blood group; kell blood group antigen; kell blood group glycoprotein; Kell blood group glycoprotein homolog; Kell blood group, metallo-endopeptidase; Kell protein; RGD1565470 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 23925 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction