missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kenpaullone, MedChemExpress
Description
Kenpaullone is a potent inhibitor of CDK1/cyclin B and GSK-3β, with IC50s of 0.4 μM and 23 nM, and also inhibits CDK2/cyclin A, CDK2/cyclin E, and CDK5/p25 with IC50s of 0.68 μM, 7.5 μM, 0.85 μM, respectively. Kenpaullone, a small molecule inhibitor of KLF4, reduces self-renewal of breast cancer stem cells and cell motility in vitro.
Specifications
Specifications
| Chemical Name or Material | Kenpaullone |
| CAS | 142273-20-9 |
| Color | Off-white to yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C16H11BrN2O |
| Quantity | 50 mg |
| Synonym | 9-Bromopaullone NSC-664704 |
| Solubility Information | DMSO : ≥ 35 mg/mL (106.97 mM) |
| SMILES | O=C1NC2=CC=CC=C2C(NC3=C4C=C(Br)C=C3)=C4C1 |
| Molecular Weight (g/mol) | 327.18 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?