missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Keratinocyte SFM (1X), without calcium chloride
GREENER_CHOICE

Product Code. 10505013
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10505013 500 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10505013 Supplier Gibco™ Supplier No. 37010022

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Keratinocyte-SFM (1X), without calcium chloride

Contains L-glutamine but no calcium chloride. Keratinocyte-SFM is supplied with prequalified human recombinant Epidermal Growth Factor 1-53 (EGF 1-53) and Bovine Pituitary Extract (BPE) in separate packaging. Components are not sold separately.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Epithelial Cells
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type Kit
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glutamine, Phenol Red
Without Additives No Calcium Chloride
Quantity 500 mL
Content And Storage • 500 mL Keratinocyte-SFM Basal Medium; store in refrigerator (2–8°C) and protect from light
• 25 mg Bovine Pituitary Extract (BPE); store in freezer (-5 to -20°C) and protect from light
• 2.5 μg EGF, Human Recombinant; store in freezer (-5 to -20°C) and protect from light

Complete Medium: store in refrigerator (2–8°C) and protect from light

Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.