missing translation for 'onlineSavingsMsg'
Learn More

Keratinocyte-SFM, with L-glutamine
GREENER_CHOICE

Product Code. 11590526 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 x 500 mL
Unit Size:
5000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11590526 10 x 500 mL 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11590526 Supplier Gibco Supplier No. 17005059
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

with L-Glutamine

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Keratinocyte Cells, Epithelial Cells
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives Glutamine, Phenol Red
Quantity 10 x 500 mL
Culture Type Mammalian Cell Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.