missing translation for 'onlineSavingsMsg'
Learn More

Kerosene 20000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30327247
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30327247 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30327247

Brand: Dr. Ehrenstorfer DREGA09010314DI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Kerosene (Unweathered)
CAS 8008-20-6
Analyte or Component Names Kerosene (Unweathered)
Shipping Condition Room Temperature
Concentration 20000 μg/mL
Certifications/Compliance ISO 17034
Synonym Kerosene, Kerosine, Kerosine (petroleum), AF 100 (pesticide), Avtur, Avtur (pesticide), Bayol 35, Bitumen Cutter, Chiotekku, Chiotekku SH, Coal oil, Conoco 170E, Deotomisol S, Eneos Kerosene, Escaid 100, Escaid 110, Exxsol D 200/240, Finalan 75, Fuels, kerosine, Ink oil, JP-A 1, KO 30 (solvent), Kedrul 212, Ketrul 211, Kixxol, Magisol 52, Neochiozol, Nysolvin 75A, Odorless Solvent 3440, P 878, Parasol, Pegasol 3040, RP 3, RP 3 (solvent), SX 12, SX 7, Shell 140, Shellsol 2046, Solbrax Eco 255/285, TS 1, W-Kerosene
Solvent or Matrix Dichloromethane
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.