missing translation for 'onlineSavingsMsg'
Learn More

Ketoprofen, MedChemExpress

Product Code. 16472099
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472099 1 g 1g
16420838 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16472099 Supplier Medchem Express Supplier No. HYB0227
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ketoprofen (RP-19583) is a non-steroidal anti-inflammatory agent. Ketoprofen can inhibits the activity of cyclooxygenase with IC50 values of 2 nM (COX-1) and 26 nM (COX-2). which is potential in the research of inflammation, immunology, and metabolic disease such as obesity.

Ketoprofen (RP-19583) is a non-steroidal anti-inflammatory agent. Ketoprofen can inhibits the activity of cyclooxygenase with IC50 values of 2 nM (COX-1) and 26 nM (COX-2). which is potential in the research of inflammation, immunology, and metabolic disease such as obesity.

Specifications

Chemical Name or Material Ketoprofen
CAS 22071-15-4
Color White
Purity Grade Notes Research
Molecular Formula C16H14O3
Quantity 1 g
Synonym RP-19583
Solubility Information DMSO : ≥ 100 mg/mL (393.27 mM) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(C1=CC=CC=C1)C2=CC=CC(C(C(O)=O)C)=C2
Molecular Weight (g/mol) 254.28
Formula Weight 254.28
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.