missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Ki-67, Rabbit Monoclonal Antibody
Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ Ki-67, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.
223.00 € - 865.00 €
Specifications
Antigen | Ki-67 |
---|---|
Clone | SP6 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12306292
|
Epredia™
RM-9106-R7 |
Prediluted | 7 mL |
794.00 €
7mL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12673697
|
Epredia™
RM-9106-RQ |
Ready-to-Use | 12 mL |
854.00 €
12mL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12603707
|
Epredia™
RM-9106-S1 |
Liquid | 500 μL, 0.5mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12683697
|
Epredia™
RM-9106-S0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12693697
|
Epredia™
RM-9106-S |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Ki-67 is a nuclear protein, which is expressed in proliferating cells. Ki-67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiescent) phase are negative for this protein.Host Species: Rabbit
Clone: SP6
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: C-terminus
Immunogen: A synthetic peptide derived from the human Ki-67 protein
Positive Control: Spleen, tonsil, or Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Ki-67 | |
Monoclonal | |
Rabbit | |
Cancer and Tumor Biology | |
Primary |
SP6 | |
Unconjugated | |
IVD | |
A synthetic peptide derived from the human Ki-67 protein |