missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki67/MKI67 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
310.00 € - 554.00 €
Specifications
| Antigen | Ki67/MKI67 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18010594
|
Novus Biologicals
NBP2-54656 |
100 μL |
554.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666858
|
Novus Biologicals
NBP2-54656-25ul |
25 μL |
310.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ki67/MKI67 Polyclonal antibody specifically detects Ki67/MKI67 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
| Ki67/MKI67 | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
| antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
| MKI67 | |
| IgG | |
| Affinity Purified | |
| 359 kDa |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| 4288 | |
| This KI67/MKI67 Antibody was developed against a Recombinant Protein corresponding to amino acids 402-55 from Human KI67/MKI67: PAKVEDAADSATKPENLSSKTRGSIPTDVEVLPTETEIHNEPFLTLWLTQVERKIQKDSLSKPEKLGTTAGQMCSGLPGLSSVDINNFGDSINESEGIPLKRRRVSFGGHLRPELFDENLPPNTPLK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title