missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA0100 Recombinant Mouse Monoclonal Antibody (A0-F3-R)
Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556502
This item is not returnable.
View return policy
Description
KIAA0100 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Belongs to the UPF0378 family. 4 isoforms of the human protein are produced by alternative splicing. Song J., Biochim. Biophys. Acta 1760:62-69(2006). Nagase T., DNA Res. 2:37-43(1995). Chen G., Leuk. Res. 29:503-509(2005).
Specifications
| KIAA0100 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q14667 | |
| KIAA0100 | |
| Recombinant protein within Human KIAA0100 aa 1,947-2,235/2,235. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| A0-F3-R | |
| Unconjugated | |
| KIAA0100 | |
| Antigen MLAA-22; BCOX; BCOX1; breast cancer overexpressed gene 1; breast cancer-overexpressed gene 1 protein; cancer/testis antigen 101; CT101; FMP27; K0100; KIAA0100; protein KIAA0100; U937-associated antigen | |
| Mouse | |
| Protein A | |
| RUO | |
| 9703 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction