Learn More
KIAA0922 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23360-1-AP-20UL
 View more versions of this product
Product Code. 18025405
  • 145.00 € / 20µL
Description
The KIAA0922 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIAA0922. This antibody recognizes human, mouse antigen. The KIAA0922 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Specifications
KIAA0922
270 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Immunofluorescence, Immunohistochemistry, ELISA
KIAA0922, TMEM131L, Transmembrane protein 131 like
20 μL
23240
Store at -20°C. Stable for one year after shipment.