missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIF11 Recombinant Rabbit Monoclonal Antibody (23GB2025), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553172
This item is not returnable.
View return policy
Description
This gene encodes a motor protein that belongs to the kinesin-like protein family. Members of this protein family are known to be involved in various kinds of spindle dynamics. The function of this gene product includes chromosome positioning, centrosome separation and establishing a bipolar spindle during cell mitosis.
Specifications
| KIF11 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| KIF11 | |
| Eg5; HKSP; KIF11; Kif8; Kifl1; Kines; kinesin 11; kinesin family member 11; kinesin-like 1; kinesin-like protein 1; Kinesin-like protein KIF11; kinesin-like spindle protein HKSP; Kinesin-related motor protein Eg5; Knsl1; MCLMR; Thyroid receptor-interacting protein 5; TR-interacting protein 5; TRIP5; TRIP-5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3832 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2025 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P52732 | |
| KIF11 | |
| A synthesized peptide derived from human Eg5 (850-1056AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction