missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF13A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 572.00 €
Specifications
| Antigen | KIF13A |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18432471
|
Novus Biologicals
NBP1-88757 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18454841
|
Novus Biologicals
NBP1-88757-25ul |
25 μL |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIF13A Polyclonal antibody specifically detects KIF13A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| KIF13A | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| bA500C11.2, FLJ27232, homolog of mouse KIF13A mannose-6-phosphate receptor transporter, kinesin family member 13A, kinesin-like protein KIF13A, Kinesin-like protein RBKIN, RBKIN | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: TGLPLNLSNFVFCQYTFWDQCESTVAAPVVDPEVPSPQSKDAQYTVTFSHCKDYVVNVTEEFLEFISDGALAIEVWGHRCAGNGSSIWEVDSLHAK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 63971 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title