Learn More
KIF15 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30774-1-AP-150UL
 View more versions of this product
Product Code. 18080696
  • 410.00 € / 150µL
Description
The KIF15 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIF15. This antibody recognizes human antigen. The KIF15 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
KIF15
700 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
FLJ25667, HKLP2, KIF15, kinesin family member 15, Kinesin like protein 2, Kinesin like protein 7, Kinesin like protein KIF15, KLP2, KNSL7, NY BR 62
150 μL
56992
Store at -20°C. Stable for one year after shipment.