Learn More
KIF1A Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24841-1-AP-20UL
 View more versions of this product
Product Code. 18054715
  • 145.00 € / 20µL
Description
The KIF1A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIF1A. This antibody recognizes human, mouse, rat antigen. The KIF1A antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
KIF1A
300 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, Immunofluorescence, ELISA
ATSV, C2orf20, HUNC 104, KIF1A, kinesin family member 1A, Kinesin like protein KIF1A, Microtubule based motor KIF1A, UNC104
20 μL
547
Store at -20°C. Stable for one year after shipment.