missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF3A Antibody (SR2233), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21780-100ul
This item is not returnable.
View return policy
Description
KIF3A Monoclonal antibody specifically detects KIF3A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| KIF3A | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR2233 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| KIF3, kinesin family member 3A, kinesin family protein 3A, kinesin-like protein KIF3A, Microtubule plus end-directed kinesin motor 3A | |
| A synthesized peptide derived from human KIF3A (Uniprot #: Q9Y496) | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience, Signal Transduction, Stem Cells | |
| 11127 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction