Learn More
KIF5C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25897-1-AP-150UL
 View more versions of this product
Product Code. 16814455
  • 410.00 € / 150µL
Description
Kinesins are a superfamily of microtubule-associated motor proteins involved in a variety of cellular processes including membranous organelle transport and cell division. Kinesin has been found in a variety of organisms and cell types and is subject to spatial and temporal regulation. These proteins have a modular structure including a conserved motor domain of approximately 350 amino acids, which is responsible for microtubule binding and ATP hydrolysis. In addition to the motor domain, subfamily members share common domain organization, exhibit sequence similarity, motility properties, and cellular functions outside of the motor domai. There are currently three known Kinesin 5 family members denoted as A, B, and C. Kinesin 5A and kinesin 5C appear to be exclusively neuronal, whereas kinesin 5B appears to be ubiquitous in its expression.
Specifications
Specifications
KIF5C
Polyclonal
Unconjugated
KIF5C
KIAA0531, KIF5C, kinesin family member 5C, Kinesin heavy chain isoform 5C, KINN, NKHC, NKHC 2, NKHC2
Rabbit
Antigen Affinity Chromatography
RUO
16574, 311024, 3800
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.79 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O60282, P28738, P56536
Kif5c
KIF5C Fusion Protein Ag23163
150 μL
Primary
Human, Mouse, Rat