missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIM-1 Recombinant Rabbit Monoclonal Antibody (309)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

109.00 € - 429.00 €

Specifications

Antigen KIM-1
Clone 309
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30534294
GREENER_CHOICE
View Documents
Invitrogen™
MA560560
100 μL
429.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30535955
View Documents
Invitrogen™
MA560561
20 μL
109.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KIM-1 Recombinant Monoclonal Antibody for ELISA

CD365 also known as T-cell immunoglobulin and mucin domain 1 (Tim-1) is a type 1 surface glycoprotein that consists of extracellular N-terminal immunoglobulin V-like domain and a mucin domain, and a short cytoplasmic tail. The endogenous ligand for CD365 is Tim-4. In addition, CD365 is known to be a cellular receptor or co-receptor for a number of viruses including hepatitis A and C viruses, Ebola virus, Marburg virus, Dengue virus, Zika virus, West Nile virus, and Japanese encephalitis virus. The direct binding of the viruses by CD365 occurs via phosphatildyl-serine exposed on viral envelopes. Polymorphisms within human CD365 gene (HAVCR1) are associated with susceptibility to infection and the disease severity. CD365 has also been implicated in the T cell co-stimulation. CD365 is primarily expressed in epithelial cells. It is also highly expressed in A549, 769-P and Caco-2 and cell lines. CD365 exists also as a soluble protein generated by metalloproteinase mediated proteolytic cleavage of the surface protein.
Specifications

Specifications

KIM-1
1 mg/mL
ELISA
Unconjugated
Rabbit
RUO
PBS with no preservative
AI503787; CD365; HAVCR; HAVCR1; HAVcr-1; Hepatitis A virus cellular receptor 1; hepatitis A virus cellular receptor 1 homolog; Kidney injury molecule 1; Kim1; KIM-1; T cell immunoglobin domain and mucin domain protein 1; t cell immunoglobulin and mucin domain-containing protein 1; T cell membrane protein 1; T-cell immunoglobulin and mucin domain containing 1; T-cell immunoglobulin and mucin domain-containing protein 1; T-cell immunoglobulin mucin family member 1; T-cell immunoglobulin mucin receptor 1; T-cell membrane protein 1; TIM; TIM1; TIM-1; TIMD1; TIMD-1
Havcr1
Primary
Protein A
309
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Canine
Havcr1
612069
Recombinant Canine TIM-1/KIM-1/HACVR protein (Tyr21-Ser160).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.