missing translation for 'onlineSavingsMsg'
Learn More

Kininogen 1 Antibody (KNG1/7422), Novus Biologicals™

Mouse Monoclonal Antibody

256.00 € - 484.00 €

Specifications

Antigen Kininogen 1
Clone KNG1/7422
Concentration 0.2 mg/ml
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30300767
View Documents Promotion Details
Novus Biologicals
NBP3-44613-20ug
20 μg
256.00 €
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30292190
View Documents Promotion Details
Novus Biologicals
NBP3-44613-100ug
100 μg
484.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Kininogen 1 Monoclonal antibody specifically detects Kininogen 1 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

Kininogen 1
0.2 mg/ml
Monoclonal
Purified
RUO
Human
Alpha-2-thiol proteinase inhibitor, BDK, BK, bradykinin, Fitzgerald factor, High molecular weight kininogen, HMWK, kininogen-1, Williams-Fitzgerald-Flaujeac factor
Recombinant fragment (around aa1-200) of human Kininogen 1 protein (Uniprot: P01042). Exact sequence is proprietary.
Primary
Store at 4⪚C.
KNG1/7422
Immunohistochemistry
Unconjugated
Mouse
Apoptosis, Immunology
10 mM PBS with 0.05% BSA
3827
IgG1 κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.