missing translation for 'onlineSavingsMsg'
Learn More

Kininogen 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

216.00 € - 326.00 €

Specifications

Antigen Kininogen 1
Dilution ELISA 1:5000-1:10000
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18659951
View Documents
Novus Biologicals
NBP2-98281-100ul
100 μg
326.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18642534
View Documents
Novus Biologicals
NBP2-98281-50ul
50 μg
216.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Kininogen 1 Polyclonal antibody specifically detects Kininogen 1 in Mouse samples. It is validated for ELISA
Specifications

Specifications

Kininogen 1
ELISA
Unconjugated
Rabbit
Apoptosis, Immunology
0.2 um filtered solution in PBS
3827
IgG
Antigen and protein A Affinity-purified
ELISA 1:5000-1:10000
Polyclonal
Purified
RUO
Mouse
Alpha-2-thiol proteinase inhibitor, BDK, BK, bradykinin, Fitzgerald factor, High molecular weight kininogen, HMWK, kininogen-1, Williams-Fitzgerald-Flaujeac factor
Produced in rabbits immunized with purified, recombinant Mouse Kininogen 1 (Accession#: NP_001095882.1; Glu 21-Ser 480)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.