missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir2.1 Antibody (SR1939), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22328-50ul
This item is not returnable.
View return policy
Description
Kir2.1 Monoclonal antibody specifically detects Kir2.1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| Kir2.1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| SR1939 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Cardiac inward rectifier potassium channel, HHBIRK1, HHIRK1, hIRK1, Inward rectifier K(+) channel Kir2.1, inward rectifier K+ channel KIR2.1, inward rectifier potassium channel 2, IRK-1, IRK1LQT7, Kir2.1, Potassium channel, inwardly rectifying subfamily J member 2, potassium inwardly-rectifying channel, subfamily J, member 2, SQT3 | |
| A synthesized peptide derived from human Kir2.1 (Uniprot #: P63252) | |
| 50 μg | |
| Neuroscience | |
| 3759 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction