missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ KIR2DL3/CD158b2 293T Cell Transient Overexpression Lysate

Product Code. 18329468 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
This item is not returnable. View return policy

Product Code. 18329468

Brand: Novus Biologicals™ H00003804T02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Overexpression Lysate

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Research Category Apoptosis
Target KIR2DL3/CD158b2
Synonym CD158 antigen-like family member B2, CD158b2 antigen, CD158B2CD158b, cl-6, killer cell immunoglobulin-like receptor 2DL3, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3, Killer inhibitory receptor cl 2-3, KIR-023GB, KIRCL23, KIR-K7b, KIR-K7c, MGC129943, MHC class I NK cell receptor, natural killer associated transcript 2, natural killer cell inhibitory receptor KIR2DL3, Natural killer-associated transcript 2, NKAT, nkat2, NKAT-2, NKAT2a, NKAT2B, NKAT2GL183, NK-receptor, p58, p58 natural killer cell receptor clone CL-6, p58 NK receptor CL-6, p58.2 MHC class-I specific NK receptor, p58.2 MHC class-I-specific NK receptor
Gene Symbol KIR2DL3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.