missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ KIR3DL2/CD158k Antibody (lacutamab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28581-100ug
This item is not returnable.
View return policy
Description
KIR3DL2/CD158k Monoclonal antibody specifically detects KIR3DL2/CD158k in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| KIR3DL2/CD158k | |
| Monoclonal | |
| Unconjugated | |
| P43630 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| lacutamab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| killer cell immunoglobulin-like receptor 3DL2, CD158K, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 2, NKAT4, NKAT-4, NKAT4B, p140 | |
| KIR3DL2/ CD158k | |
| 100 μg | |
| Immunology | |
| 3812 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction