missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR3DL3/CD158z Antibody, R&D Systems™

Rabbit Monoclonal Antibody
209.00 € - 439.00 €
Specifications
| Antigen | KIR3DL3/CD158z |
|---|---|
| Clone | 1136B |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18795119
![]() |
R&D Systems
MAB8919-SP |
25 μg |
209.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18758528
![]() |
R&D Systems
MAB8919 |
100 μg |
439.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIR3DL3/CD158z Monoclonal specifically detects KIR3DL3/CD158z in Human samples. It is validated for Flow Cytometry.Specifications
| KIR3DL3/CD158z | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| 115653 | |
| Human embryonic kidney cell line HEK293-derived recombinant human KIR3DL3/CD158z Met1-Leu322 Accession # Q8N743 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human KIR3DL3/CD158z in direct ELISAs. |
| 1136B | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8N743 | |
| KIR3DL3 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title