missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kirrel3/NEPH2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
241.00 € - 326.00 €
Specifications
| Antigen | Kirrel3/NEPH2 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18665922
|
Novus Biologicals
NBP2-98153-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603242
|
Novus Biologicals
NBP2-98153-50ul |
50 μg |
241.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kirrel3/NEPH2 Polyclonal antibody specifically detects Kirrel3/NEPH2 in Rat samples. It is validated for ELISASpecifications
| Kirrel3/NEPH2 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| 0.2 um filtered solution in PBS | |
| 84623 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| KIAA1867MGC126824, kin of IRRE like 3 (Drosophila), Kin of irregular chiasm-like protein 3, kin of IRRE-like protein 3, KIRREPRO4502, MRD4, NEPH2MGC126850, nephrin-like 2, Nephrin-like protein 2 | |
| Produced in rabbits immunized with purified, recombinant Rat Kirrel3/NEPH2 (Uniprot#: Q09GS6; Met1-Ala523). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title