missing translation for 'onlineSavingsMsg'
Learn More

Velp™ KjTabs VCT - Kjeldahl Catalyst Tablets, Velp™

Product Code. 31781791 Shop All Velp Products
Change view
Click to view available options
Quantity:
1000/Pk., 1000 Pc./Box
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31781791 1000/Pk., 1000 Pc./Box Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31781791 Supplier Velp™ Supplier No. A00000276
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

KjTabsTM VCT Kjeldahl Catalyst Tablets are ideal for tougher samples. The perfect solutions to speed up the digestion process.

  • The addition of a suitable catalyst, along with high temperatures, influences the speed and the efficiency of mineralization in Kjeldahl analyses.
  • Achieving the highest level of performance in a wide range of Kjeldahl applications is now possible with genuine catalyst tablets.
  • The use of catalysts is now recognized and officially accepted by the major standards such as AOCS, AOAC, AACC and ISO to accelerate Kjeldahl digestion.
  • KjTabs™ VCM consist of accurately pre-dosed tablets composed of sulfate, to increase the boiling point of sulphuric acid, plus Copper (Cu) as metal salt to accelerate the mineralization process.
  • Universally applicable, with a low copper content 3.5g K2SO4 + 0.1g CuSO4 x 5H2O 1000 pcs.
  • Most suitable choice when performing Kjeldahl analysis in a great variety of industries such as food and beverage, agriculture, environmental, chemical and petrochemical:
    • Protein determination in Nuts and Nut Products (almonds, nuts, hazelnuts, coconuts), grains and cereals
    • N/Protein determination in flour (soya beans and lupins)
    • Crude protein determination in meat and meat products
    • Protein determination in compressed and granular yeast
    • Protein determination in sugar, syrup, molasses
    • Protein determination in wheat spaghetti and macaroni, egg pasta
    • Nitrogen content determination in urea
    • Total nitrogen determination in water
    • Nitrogen content determination in raw natural rubber and rubber latex

Specifications

Chemical Name or Material VCT
Color White
Quantity 1000/Pk., 1000 Pc./Box
Packaging Metal Tin
Grade Reagent
compliance-icons compliance-icons compliance-icons
Product Identifier
  • KJTABS VCT
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH212-Warning! Hazardous respirable dust may be formed when used. Do not breathe dust.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.