missing translation for 'onlineSavingsMsg'
Learn More

KKL-35, MedChemExpress

Product Code. 16486347
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486347 5 mg 5mg
16480929 1 mg 1mg
16413628 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486347 Supplier Medchem Express Supplier No. HY101866
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress KKL-35 is a trans-translation tagging reaction inhibitor with an IC50 of 0.9 μM.

KKL-35 is a trans-translation tagging reaction inhibitor with an IC50 of 0.9 μM.

Specifications

Chemical Name or Material KKL-35
CAS 865285-29-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H9ClFN3O2
Quantity 5 mg
Solubility Information DMSO : 6.25 mg/mL (19.67 mM; Need ultrasonic)
SMILES O=C(NC1=NN=C(C2=CC=C(F)C=C2)O1)C3=CC=C(Cl)C=C3
Molecular Weight (g/mol) 317.7
Formula Weight 317.7
Percent Purity 95.88%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.