missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KLC1 Recombinant Rabbit Monoclonal Antibody (24GB6030)

Rabbit Recombinant Monoclonal Antibody
198.00 € - 564.00 €
Specifications
| Antigen | KLC1 |
|---|---|
| Clone | 24GB6030 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275932
![]() |
Invitrogen™
MA557432 |
100 μL |
564.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275689
|
Invitrogen™
MA557433 |
20 μL |
198.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Conventional kinesin is a tetrameric molecule composed of two heavy chains and two light chains, and transports various cargos along microtubules toward their plus ends. The heavy chains provide the motor activity, while the light chains bind to various cargos. This gene encodes a member of the kinesin light chain family. It associates with kinesin heavy chain through an N-terminal domain, and six tetratricopeptide repeat (TPR) motifs are thought to be involved in binding of cargos such as vesicles, mitochondria, and the Golgi complex. Thus, kinesin light chains function as adapter molecules and not motors per se. Although previously named 'kinesin 2', this gene is not a member of the kinesin-2 / kinesin heavy chain subfamily of kinesin motor proteins. Extensive alternative splicing produces isoforms with different C-termini that are proposed to bind to different cargos; however, the full-length nature of some of these variants has not been determined.Specifications
| KLC1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| KLC1 | |
| AI874768; kinesin 2; kinesin 2 60/70kDa; kinesin light chain 1; Kinesin2; Klc; KLC 1; Klc1; KNS 2; KNS 2A; Kns2; KNS2A; medulloblastoma antigen MU-MB-2.50; MGC15245 | |
| KLC1 | |
| Primary | |
| Affinity chromatography |
| 24GB6030 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88447, P37285, Q07866 | |
| 16593, 171041, 3831 | |
| A synthesized peptide derived from human KLC1 (aa 501-573). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title